Potassium-tert-pentyloxide 25% (w/w) in toluene
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Potassium tert-amylate, Potassium 2-methyl-2-butoxide, Potassium tert-pentoxide
L15111-AU
L15111-AK
AAAL15111-AUEA
262
USD
AAAL15111-AU
AAAL15111-AK
Potassium-tert-pentyloxide 25% (w/w) in toluene
Potassium-tert-pentyloxide
Fieser: 1,939 9,379
CAS No.: 41233-93-6
Molecular Formula: C5H11KO
Formula Weight: 126.24
Flash Point: 4°(39°F)
Beilstein Reference: 4149025
UN No.: 2924
MDL No.: MFCD00064808
Formula:
C₅H₁₁KO Density: 0.870 Flash Pt: 4 °C(39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064808 CAS Number: 41233-93-6 EINECS: 255-272-5 UN: 2924 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...