Lithiumbis(trimethylsilyl)amide 20% ∼1.06 M in THF/ethylbenzene

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt

L15012-AE
AAAL15012-AEEA 101 USD
AAAL15012-AE
Lithiumbis(trimethylsilyl)amide 20% ∼1.06 M in THF/ethylbenzene
Lithium bis(trimethylsilyl)amide

Packaged in resealable septum cap bottle


CAS No.: 4039-32-1 Molecular Formula: C6H18LiNSi2 Formula Weight: 167.33 Flash Point: -17°(1°F) Beilstein Reference: 8136861 UN No.: 2924 MDL No.: MFCD00008261

Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
Density: 0.891
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008261
CAS Number: 4039-32-1
EINECS: 223-725-6
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR