5-Amino-2-chlorobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

L14501-14 L14501-22
AAAL14501-14EA 50.4 USD
AAAL14501-14 AAAL14501-22
5-Amino-2-chlorobenzoic acid 98+%
5-Amino-2-chlorobenzoic acid

Purity: 98+% CAS No.: 89-54-3 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: ca 185° dec. Beilstein Reference: 2090422

Formula: C₇H₆ClNO₂
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: ca 185 °C (dec.)
Density: 1.5
Storage Temperature: Ambient
MDL Number: MFCD00007845
CAS Number: 89-54-3
EINECS: 201-916-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR