Triethylammonium dihydrogentrifluoride (∼37% HF)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride

L14417-06 L14417-14 L14417-22
AAAL14417-06EA 29.12 USD
AAAL14417-06 AAAL14417-14 AAAL14417-22
Triethylammonium dihydrogentrifluoride (∼37% HF)
Triethylammonium dihydrogentrifluoride

CAS No.: 73602-61-6 Molecular Formula: C6H15N.3HF Formula Weight: 161.21 Flash Point: 87°(188°F) Refractive Index: 1.3915 Melting Point: -29° Boiling Point: 70°/15mm Beilstein Reference: 5522945 UN No.: 1790 MDL No.: MFCD00043294

Formula: C₆H₁₈F₃N
MW: 161.21
Boiling Pt: 70 °C/15 mm
Melting Pt: –29 °C
Density: 0.990
Flash Pt: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043294
CAS Number: 73602-61-6
EINECS: 277-550-5
UN: 1790
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR