Dowex® 1X8 (Cl), Dowex® 100-200 mesh

Supplier: Thermo Scientific Chemicals

L14257-18 L14257-30 L14257-0B
AAAL14257-18EA 46.7 USD
AAAL14257-18 AAAL14257-30 AAAL14257-0B
Dowex® 1X8 (Cl), Dowex® 100-200 mesh
Dowex® 1X8 (Cl)

CAS No.: 12627-85-9 Odor: Amine-like MDL No.: MFCD00132718

Storage Temperature: Ambient
MDL Number: MFCD00132718
CAS Number: 12627-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR