Luteolin 97%
Supplier: Thermo Scientific Chemicals
L14186-MC
L14186-ME
AAAL14186-MCEA
76.9
USD
AAAL14186-MC
AAAL14186-ME
Luteolin 97%
Luteolin
An apoptosis inducer
Purity: 97%
CAS No.: 491-70-3
Molecular Formula: C15H10O6
Formula Weight: 286.24
Melting Point:
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...