Dowex® 50WX2 (H), Dowex® 200-400 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13945-0B
L13945-18
L13945-30
AAAL13945-0BEA
840
USD
AAAL13945-0B
AAAL13945-18
AAAL13945-30
Dowex® 50WX2 (H), Dowex® 200-400 mesh
Dowex® 50WX2 (H)
CAS No.: 12612-37-2
MDL No.: MFCD00132722
Storage Temperature:
Ambient |
MDL Number:
MFCD00132722 CAS Number: 12612-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...