L-α-Aminoadipic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid
L13924-03
L13924-06
AAAL13924-03EA
99.4
USD
AAAL13924-03
AAAL13924-06
L-α-Aminoadipic acid 98%
L-α-Aminoadipic acid
Purity: 98%
CAS No.: 1118-90-7
Molecular Formula: C6H11NO4
Formula Weight: 161.16
Melting Point: ca 206° dec.
Beilstein Reference: 1724348
Formula:
C₆H₁₁NO₄ MW: 161.16 g/mol Melting Pt: ∼ 206 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002636 CAS Number: 1118-90-7 |
Specification Test Results
Optical rotation | +25° (c=2 in 5N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...