Dowex® 50WX8 (H), Dowex® 200-400 mesh

Supplier: Thermo Scientific Chemicals
Danger

L13922-0B L13922-18 L13922-30
AAAL13922-0BEA 682 USD
AAAL13922-0B AAAL13922-18 AAAL13922-30
Dowex® 50WX8 (H), Dowex® 200-400 mesh
Dowex® 50WX8 (H)

CAS No.: 11119-67-8 MDL No.: MFCD00132726

Storage Temperature: Ambient
MDL Number: MFCD00132726
CAS Number: 11119-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR