N-Methylaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13873-0E
L13873-22
L13873-36
AAAL13873-0EEA
152
USD
AAAL13873-0E
AAAL13873-22
AAAL13873-36
N-Methylaniline 98%
N-Methylaniline
Purity: 98%
CAS No.: 100-61-8
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 86°(186°F)
Refractive Index: 1.5710
Melting Point: -57°
Boiling Point: 194-197°
Beilstein Reference: 741982
Merck Reference: 14,6019
UN No.: 2294
MDL No.: MFCD00008283
Formula:
C₇H₉N Boiling Pt: 194…197 °C Melting Pt: –57 °C Density: 0.989 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008283 CAS Number: 100-61-8 EINECS: 202-870-9 UN: 2294 ADR: 6.1,III Merck Index: 14,6019 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...