N-Methylaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

L13873-0E L13873-22 L13873-36
AAAL13873-0EEA 152 USD
AAAL13873-0E AAAL13873-22 AAAL13873-36
N-Methylaniline 98%
N-Methylaniline

Purity: 98% CAS No.: 100-61-8 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 86°(186°F) Refractive Index: 1.5710 Melting Point: -57° Boiling Point: 194-197° Beilstein Reference: 741982 Merck Reference: 14,6019 UN No.: 2294 MDL No.: MFCD00008283

Formula: C₇H₉N
Boiling Pt: 194…197 °C
Melting Pt: –57 °C
Density: 0.989
Flash Pt: 86 °C (186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008283
CAS Number: 100-61-8
EINECS: 202-870-9
UN: 2294
ADR: 6.1,III
Merck Index: 14,6019

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR