N-Butylethylenediamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Amino-2-(butylamino)ethane, N-Butyl-1,2-diaminoethane
L13437-06
L13437-14
AAAL13437-06EA
100.58
USD
AAAL13437-06
AAAL13437-14
N-Butylethylenediamine 97%
N-Butylethylenediamine
Purity: 97%
CAS No.: 19522-69-1
Molecular Formula: CH3(CH2)3NHCH2CH2NH2
Formula Weight: 116.21
Flash Point: 59°(138°F)
Refractive Index: 1.4430
Boiling Point: 170-171°
Beilstein Reference: 1732700
UN No.: 2734
MDL No.: MFCD00025602
Formula:
C₆H₁₆N₂ MW: 116.21 g/mol Boiling Pt: 71 Density: 0.836 Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025602 CAS Number: 19522-69-1 UN: 2734 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...