6-Fluoro-m-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Amino-4-fluorotoluene, 6-Fluoro-m-methylaniline, 2-Fluoro-5-methylaniline
L13411-06
L13411-14
AAAL13411-06EA
24.2
USD
AAAL13411-06
AAAL13411-14
6-Fluoro-m-toluidine 98%
6-Fluoro-m-toluidine
Purity: 98%
CAS No.: 452-84-6
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 81°(177°F)
Refractive Index: 1.5330
Odor: Characteristic
Boiling Point: 80-86°/11mm
Beilstein Reference: 2715995
UN No.: 2810
MDL No.: MFCD00007654
Formula:
C₇H₈FN MW: 125.15 g/mol Boiling Pt: 80…86 °C (11 mmHg) Density: 1.109 Flash Pt: 81 °C (177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007654 CAS Number: 452-84-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...