6-Nitro-m-anisic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Methoxy-2-nitrobenzoic acid
L13346-06
L13346-14
AAAL13346-06EA
22.6
USD
AAAL13346-06
AAAL13346-14
6-Nitro-m-anisic acid 97%
6-Nitro-m-anisic acid
Purity: 97%
CAS No.: 1882-69-5
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 130-135°
Beilstein Reference: 1877657
Formula:
C₈H₇NO₅ MW: 197.15 g/mol Melting Pt: 138 Storage Temperature: Ambient |
MDL Number:
MFCD00151836 CAS Number: 1882-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...