2-Pentanone 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl Propyl Ketone

L13262-22 L13262-36 L13262-14
AAAL13262-22EA 43.67 USD
AAAL13262-22 AAAL13262-36 AAAL13262-14
2-Pentanone 99+%
2-Pentanone

Purity: 99+% CAS No.: 107-87-9 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 7°(44°F) Refractive Index: 1.3905 Melting Point: -78° Boiling Point: 100-101° Beilstein Reference: 506058 Merck Reference: 14,6114 UN No.: 1249 MDL No.: MFCD00009400

Formula: C₅H₁₀O
Boiling Pt: 100…101 °C
Melting Pt: –78 °C
Density: 0.813
Flash Pt: 7 °C (44 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009400
CAS Number: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3,II
Merck Index: 14,6114

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR