N-Octyl-β-D-glucopyranoside
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside
L13259-03
L13259-MD
AAAL13259-03EA
106
USD
AAAL13259-03
AAAL13259-MD
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
Non-ionic detergent for membrane proteins
CAS No.: 29836-26-8
Molecular Formula: C14H28O6
Formula Weight: 292.37
Melting Point: 103-104°
Beilstein Reference: 84118
Optical Rotation: -31° (c=2 in methanol)
Formula:
C₁₄H₂₈O₆ MW: 292.37 g/mol Melting Pt: 110 Storage Temperature: Ambient |
MDL Number:
MFCD00063288 CAS Number: 29836-26-8 EINECS: 249-887-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...