Calcium chloride, dried 97%, powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13191-30
L13191-0B
L13191-0I
AAAL13191-30EA
45.27
USD
AAAL13191-30
AAAL13191-0B
AAAL13191-0I
Calcium chloride, dried 97%, powder
Calcium chloride
Purity: 97%
CAS No.: 10043-52-4
Molecular Formula: CaCl2
Formula Weight: 110.99
Melting Point: 782°
Boiling Point:
Formula:
CaCl₂ Boiling Pt: >1600 °C Melting Pt: 782 °C Density: 2.174 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 REACH: 01-2119494219-28 Merck Index: 14,1659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...