Sodium ethoxide 21% (w/w) in ethanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13083-0F
L13083-AP
L13083-AE
AAAL13083-0FEA
250.7
USD
AAAL13083-0F
AAAL13083-AP
AAAL13083-AE
Sodium ethoxide 21% (w/w) in ethanol
Sodium ethoxide
CAS No.: 141-52-6
Molecular Formula: C2H5NaO
Formula Weight: 68.05
Flash Point: 20°(68°F)
Refractive Index: 1.3875
Beilstein Reference: 3593646
Merck Reference: 14,8613
UN No.: 3274
MDL No.: MFCD00012417
Formula:
C₂H₅NaO Density: 0.868 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00012417 CAS Number: 141-52-6 EINECS: 205-487-5 UN: 2924 ADR: 3,III Merck Index: 14,8613 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...