4-n-Butoxyphenyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Warning

L12877-03 L12877-06
AAAL12877-03EA 35.2 USD
AAAL12877-03 AAAL12877-06
4-n-Butoxyphenyl isocyanate 98%
4-n-Butoxyphenyl isocyanate

Purity: 98% CAS No.: 28439-86-3 Molecular Formula: C11H13NO2 Formula Weight: 191.23 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5220 Boiling Point: 149-150°/3mm Beilstein Reference: 2107851 UN No.: 2206 MDL No.: MFCD00037042

Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
Boiling Pt: 149…150 °C (3 mmHg)
Density: 1.055
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00037042
CAS Number: 28439-86-3
UN: 2206
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR