1,2-Ethanedithiol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1.2-Ethanedithiol, 1,2-Dimercaptoethane
L12865-36
L12865-14
L12865-22
AAAL12865-36EA
337
USD
AAAL12865-36
AAAL12865-14
AAAL12865-22
1,2-Ethanedithiol 98+%
1,2-Ethanedithiol
Purity: 98+%
CAS No.: 540-63-6
Molecular Formula: C2H6S2
Formula Weight: 94.20
Flash Point: 45°(113°F)
Refractive Index: 1.5580
Odor: Stench
Melting Point: -41°
Boiling Point: 144-146°
Beilstein Reference: 505946
Merck Reference: 14,3725
UN No.: 3071
MDL No.: MFCD00004892
Formula:
C₂H₆S₂ MW: 94.2 g/mol Boiling Pt: 144…146 °C Melting Pt: -41 Density: 1.123 Flash Pt: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004892 CAS Number: 540-63-6 EINECS: 208-752-3 UN: 3071 ADR: 6.1 II Merck Index: 14,3725 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...