3-(4-Morpholino)propyl isothiocyanate 96%

Supplier: Thermo Scientific Chemicals
Danger

L12819-03 L12819-06
AAAL12819-03EA 28.3 USD
AAAL12819-03 AAAL12819-06
3-(4-Morpholino)propyl isothiocyanate 96%
3-(4-Morpholino)propyl isothiocyanate

Purity: 96% CAS No.: 32813-50-6 Molecular Formula: C8H14N2OS Formula Weight: 186.28 Storage Temperature: 2°C to 8°C Refractive Index: 1.5305 Boiling Point: 182-183°/13mm Beilstein Reference: 879217 UN No.: 2922 MDL No.: MFCD00041222

Formula: C₈H₁₄N₂OS
MW: 186.28 g/mol
Boiling Pt: 182…183 °C (13 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00041222
CAS Number: 32813-50-6
UN: 2922
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR