Indene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Indene

L12665-06 L12665-14 L12665-22
AAAL12665-06EA 54.4 USD
AAAL12665-06 AAAL12665-14 AAL12665-22
Indene 97%
Indene

Purity: 97% CAS No.: 95-13-6 Molecular Formula: C9H8 Formula Weight: 116.16 Storage Temperature: 2°C to 8°C Flash Point: 58°(136°F) Refractive Index: 1.5740 Odor: Aromatic Melting Point: ca -2° Boiling Point: 181-182° Beilstein Reference: 635873 Merck Reference: 14,4939 UN No.: 3295 MDL No.: MFCD00003777

Formula: C₉H₈
Boiling Pt: 181…182 °C
Melting Pt: ca …2 °C
Density: 0.996
Flash Pt: 58 °C(136 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003777
CAS Number: 95-13-6
EINECS: 202-393-6
UN: 3295
ADR: "3",III
Merck Index: 14,4939

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR