1-(4-Hydroxyphenyl)-2-thiourea 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(4-Hydroxyphenyl)thiourea, 4-Hydroxyphenylthiourea
L12537-06
L12537-03
AAAL12537-06EA
128
USD
AAAL12537-06
AAAL12537-03
1-(4-Hydroxyphenyl)-2-thiourea 98+%
1-(4-Hydroxyphenyl)-2-thiourea
Purity: 98+%
CAS No.: 1520-27-0
Molecular Formula: C7H8N2OS
Formula Weight: 168.22
Odor: Odorless
Melting Point: ca 210° dec.
Beilstein Reference: 1102429
MDL No.: MFCD00041186
Formula:
(C₆H₄(OH))NC(S)NH₂ MW: 168.22 g/mol Melting Pt: ∼ 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00041186 CAS Number: 1520-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...