1H,1H,5H-Octafluoropentyl methacrylate 98% stabilised
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Methacrylic Acid 1H,1H,5H-Perfluoropentyl Ester, Methacrylic Acid 1H,1H,5H-Octafluoropentyl Ester, 1H,1H,5H-Perfluoropentyl Methacrylate, 2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate
L12044-14
L12044-06
AAAL12044-14EA
284
USD
AAAL12044-14
AAAL12044-06
1H,1H,5H-Octafluoropentyl methacrylate 98% stabilised
1H,1H,5H-Octafluoropentyl methacrylate
Purity: 98%
CAS No.: 355-93-1
Molecular Formula: C9H8F8O2
Formula Weight: 300.15
Flash Point: 81°(177°F)
Refractive Index: 1.3600
Boiling Point: 178-179°
Beilstein Reference: 1801409
MDL No.: MFCD00039278
Formula:
C₉H₈F₈O₂ MW: 300.15 g/mol Boiling Pt: 178…179 °C Density: 1.432 Flash Pt: 81 °C (177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039278 CAS Number: 355-93-1 EINECS: 206-596-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...