2-Fluoro-1-methylpyridinium-p-toluenesulfonate tech. 90%, tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulfonate, 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate

L11088-03
AAAL11088-03EA 38.5 USD
AAAL11088-03 AAAL11088-06
2-Fluoro-1-methylpyridinium-p-toluenesulfonate tech. 90%, tech.
2-Fluoro-1-methylpyridinium p-toluenesulphonate

Purity: 90% CAS No.: 58086-67-2 Molecular Formula: C13H14FNO3S Formula Weight: 283.32 Melting Point: 128-131° Beilstein Reference: 4345357

Formula: C₁₃H₁₄FNO₃S
MW: 283.32 g/mol
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00011983
CAS Number: 58086-67-2
EINECS: 261-108-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR