Amyl acetate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate

L10852-22 L10852-36
AAAL10852-22EA 27.79 USD
AAAL10852-22 AAAL10852-36
Amyl acetate 99%
Amyl acetate

Purity: 99% CAS No.: 628-63-7 Molecular Formula: CH3CO2(CH2)4CH3 Formula Weight: 130.19 Flash Point: 24°(75°F) Refractive Index: 1.4030 Odor: Fruit-like Melting Point: -100° Boiling Point: 142° Beilstein Reference: 1744753 UN No.: 1104 MDL No.: MFCD00009500

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 149
Melting Pt: -71
Density: 0.876
Flash Pt: 24 °C (75 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009500
CAS Number: 628-63-7
EINECS: 211-047-3
UN: 1104
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37af9fce9b4d1452153c5c88d6e47d0d5e08f221&vid=b0e4dd5050dab83dda2db704a160c2058b4bd15a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR