Amyl acetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
L10852-22
L10852-36
AAAL10852-22EA
27.79
USD
AAAL10852-22
AAAL10852-36
Amyl acetate 99%
Amyl acetate
Purity: 99%
CAS No.: 628-63-7
Molecular Formula: CH3CO2(CH2)4CH3
Formula Weight: 130.19
Flash Point: 24°(75°F)
Refractive Index: 1.4030
Odor: Fruit-like
Melting Point: -100°
Boiling Point: 142°
Beilstein Reference: 1744753
UN No.: 1104
MDL No.: MFCD00009500
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 149 Melting Pt: -71 Density: 0.876 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009500 CAS Number: 628-63-7 EINECS: 211-047-3 UN: 1104 ADR: 3 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...