3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Pyridyl)-5,6-bis(4-sulfophenyl)-1,2,4-triazine disodium salt hydrate, 3-(2-Pyridyl)-5,6-bis(4-sulphophenyl)-1,2,4-triazine disodium salt hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulphonic acid disodium salt hydrate, PDT disulphonate hydrate, FerroZine(TM) Iron reagent disodium salt hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, PDT disulfonate hydrate

L10607-06 L10607-03
AAAL10607-06EA 107 USD
AAAL10607-06 AAAL10607-03
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 98%
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate

Purity: 98% CAS No.: 28048-33-1 Molecular Formula: C20H12N4Na2O6S2.xH2O Formula Weight: 514.46(anhy) Beilstein Reference: 5222087 MDL No.: MFCD00717621

Formula: C₂₀H₁₂N₄Na₂O₆S₂·xH₂O
MW: 514.44 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00717621
CAS Number: 28048-33-1
EINECS: 248-797-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR