Perfluorokerosene, low boiling
Supplier: Thermo Scientific Chemicals
Synonyms:
PFK
L10538-03
AAAL10538-MDEA
49.51
USD
AAAL10538-MD
AAAL10538-03
Perfluorokerosene, low boiling
Perfluorokerosene
Boiling Point: ca 40-100°
MDL No.: MFCD00132835
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...