Thiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 99%
CAS No.: 288-47-1
Molecular Formula: C3H3NS
Formula Weight: 85.13
Flash Point: 22°(71°F)
Refractive Index: 1.5390
Odor: Unpleasant
Melting Point: -33°
Boiling Point: 117-118°
Beilstein Reference: 103852
Merck Reference: 14,9307
UN No.: 1993
MDL No.: MFCD00005315
Formula:
C₃H₃NS Boiling Pt: 117…118 °C Melting Pt: –33 °C Density: 1.200 Flash Pt: 22 °C (71 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005315 CAS Number: 288-47-1 EINECS: 206-021-3 UN: 1993 ADR: 3,III Merck Index: 14,9307 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...