2,2'-Butyliminodiethanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 98%
CAS No.: 102-79-4
Molecular Formula: CH3(CH2)3N(CH2CH2OH)2
Formula Weight: 161.25
Flash Point: 143°(289°F)
Refractive Index: 1.4640
Melting Point: -70°
Boiling Point: 268-272°
Beilstein Reference: 1739642
UN No.: 2735
MDL No.: MFCD00002856
Formula:
H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂ Boiling Pt: 268…272 °C Melting Pt: –70 °C Density: 0.968 Flash Pt: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002856 CAS Number: 102-79-4 EINECS: 203-055-0 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...