Guanidinium nitrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09643-0B
L09643-30
AAAL09643-0BEA
93.1
USD
AAAL09643-0B
AAAL09643-30
Guanidinium nitrate 98%
Guanidinium nitrate
Purity: 98%
CAS No.: 506-93-4
Molecular Formula: CH6N4O3
Formula Weight: 122.08
Odor: Odorless
pH: 4.9
Melting Point: 213-216°
Beilstein Reference: 3596600
Merck Reference: 14,4562
UN No.: 1467
MDL No.: MFCD00013028
Formula:
CH₆N₄O₃ Melting Pt: 213…216 °C Density: 1.44 Storage Temperature: Ambient |
MDL Number:
MFCD00013028 CAS Number: 506-93-4 EINECS: 208-060-1 UN: 1467 ADR: 5.1,III Merck Index: 14,4562 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...