2-Chloro-5-methoxybenzimidazole 94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-5-methoxy-1H-benzo[d]imidazole, 2-Chloro-5-methoxy-1H-benzimidazole

L08440-MD L08440-03
AAAL08440-MDEA 67.2 USD
AAAL08440-MD AAAL08440-03
2-Chloro-5-methoxybenzimidazole 94%
2-Chloro-5-methoxybenzimidazole

Purity: 94% CAS No.: 15965-54-5 Molecular Formula: C8H7ClN2O Formula Weight: 182.61 Melting Point: 165-168° Beilstein Reference: 777455 MDL No.: MFCD00051945

Formula: C₈H₇ClN₂O
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00051945
CAS Number: 15965-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR