Linoleic acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
L07949-06
L07949-14
L07949-22
AAAL07949-06EA
65.9
USD
AAAL07949-06
AAAL07949-14
AAAL07949-22
Linoleic acid 95%
Linoleic acid
Unsaturated omega-6 fatty acid
Purity: 95%
CAS No.: 60-33-3
Molecular Formula: C18H32O2
Formula Weight: 280.45
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₈H₃₂O₂ MW: 280.45 g/mol Boiling Pt: 360 Melting Pt: -5 Density: 0.902 Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064241 CAS Number: 60-33-3 EINECS: 200-470-9 Merck Index: 14,5505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...