4-Bromopyridinium chloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

L07052-06 L07052-14
AAAL07052-06EA 105 USD
AAAL07052-06 AAAL07052-14 AAAL07052-03
4-Bromopyridinium chloride 99%
4-Bromopyridinium chloride

Purity: 99% CAS No.: 19524-06-2 Molecular Formula: C5H4BrN.HCl Formula Weight: 194.46 Melting Point: ca 270° dec. Beilstein Reference: 3621847

Formula: C₅H₅BrClN
Melting Pt: ∼ 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR