9-Bromoanthracene 96%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06733-06
AAAL06733-06EA
36.8
USD
AAAL06733-06
AAAL06733-03
9-Bromoanthracene 96%, Technical Grade
9-Bromoanthracene
Purity: 96%
CAS No.: 1564-64-3
Molecular Formula: C14H9Br
Formula Weight: 257.14
Melting Point: 97-103°
Beilstein Reference: 1869747
Formula:
C₁₄H₉Br Boiling Pt: 359 °C (1013 hPa) Melting Pt: 97…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001243 CAS Number: 1564-64-3 EINECS: 216-359-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...