Sodium ethoxide 96%

Supplier: Thermo Scientific Chemicals
Danger

L06230-0P L06230-36 L06230-22
AAAL06230-0PEA 273 USD
AAAL06230-0P AAAL06230-36 AAAL06230-22
Sodium ethoxide 96%
Sodium ethoxide

Purity: 96% CAS No.: 141-52-6 Molecular Formula: NaOCH2CH3 Formula Weight: 68.05 Flash Point: 30°(86°F) Odor: Irritating pH: 13 Melting Point: 300° Beilstein Reference: 3593646 Merck Reference: 14,8613 UN No.: 3206 MDL No.: MFCD00012417

Formula: C₂H₅NaO
Melting Pt: >300 °C
Density: 0.868 g/cm³ (20 °C)
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3095
ADR: 8,II
Merck Index: 14,8613

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR