Sodium ethoxide 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 96%
CAS No.: 141-52-6
Molecular Formula: NaOCH2CH3
Formula Weight: 68.05
Flash Point: 30°(86°F)
Odor: Irritating
pH: 13
Melting Point:
Formula:
C₂H₅NaO Melting Pt: >300 °C Density: 0.868 g/cm³ (20 °C) Flash Pt: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00012417 CAS Number: 141-52-6 EINECS: 205-487-5 UN: 3095 ADR: 8,II Merck Index: 14,8613 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...