Potassium tetrakis(4-chlorophenyl)borate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassiumtetrakis(4-chlorophenyl)borate
L05609-03
L05609-MD
AAAL05609-03EA
162
USD
AAAL05609-03
AAAL05609-MD
Potassium tetrakis(4-chlorophenyl)borate 98%
Potassium tetrakis(4-chlorophenyl)borate
Purity: 98%
CAS No.: 14680-77-4
Molecular Formula: C24H16BCl4K
Formula Weight: 496.12
Melting Point:
Formula:
C₂₄H₁₆BCl₄K Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043105 CAS Number: 14680-77-4 EINECS: 238-723-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...