Molecular sieve A4 (0.4 nm, 4 Å), beads 0.4-0.8 mm (0.02-0.03 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate

L05512-0B L05512-30
AAAL05512-0BEA 154.53 USD
AAAL05512-0B AAAL05512-30
Molecular sieve A4 (0.4 nm, 4 Å), beads 0.4-0.8 mm (0.02-0.03 in)
Molecular sieve A4 (0.4 nm, 4 Å)

CAS No.: 70955-01-0 MDL No.: MFCD00131613

Density: 1.1…2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR