Molecular sieve A4 (0.4 nm, 4 Å), beads 3-5 mm (0.12-0.20 in)

Supplier: Thermo Scientific Chemicals

L05466-0B L05466-30 L05466-0I
AAAL05466-0BEA 148.44 USD
AAAL05466-0B AAAL05466-30 AAAL05466-0I
Molecular sieve A4 (0.4 nm, 4 Å), beads 3-5 mm (0.12-0.20 in)
Molecular sieve A4 (0.4 nm, 4 Å)
Density: 1.1…2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR