Molecular sieve A4 (0.4 nm, 4 Å), beads 1-2 mm (0.04-0.08 in)
Supplier: Thermo Scientific Chemicals
L05454-0B
L05454-0I
L05454-30
AAAL05454-0BEA
116
USD
AAAL05454-0B
AAAL05454-0I
AAAL05454-30
Molecular sieve A4 (0.4 nm, 4 Å), beads 1-2 mm (0.04-0.08 in)
Molecular sieve A4 (0.4 nm, 4 Å)
CAS No.: 70955-01-0
MDL No.: MFCD00131613
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...