Triethanolamine (trolamine) 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol
L04486-0E
L04486-36
L04486-22
AAAL04486-0EEA
123
USD
AAAL04486-0E
AAAL04486-36
AAAL04486-22
Triethanolamine (trolamine) 98+%
Triethanolamine (Trolamine)
Protein purification reagent
Purity: 98+%
CAS No.: 102-71-6
Molecular Formula: C6H15NO3
Formula Weight: 149.19
Flash Point: 185°(365°F)
Refractive Index: 1.4850
Odor: Ammonia-like
Melting Point: 19-21°
Boiling Point: 206-207°/15mm
Beilstein Reference: 1699263
Merck Reference: 14,9665
MDL No.: MFCD00002855
Formula:
C₆H₁₅NO₃ Boiling Pt: 206…207 °C/15 mm Melting Pt: 19…21 °C Density: 1.124 Flash Pt: 185 °C(365 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002855 CAS Number: 102-71-6 EINECS: 203-049-8 Merck Index: 14,9665 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...