4-Hydroxythiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04429-14
L04429-06
AAAL04429-14EA
170
USD
AAAL04429-14
AAAL04429-06
4-Hydroxythiophenol 97%
4-Hydroxythiophenol
Purity: 97%
CAS No.: 637-89-8
Molecular Formula: C6H6OS
Formula Weight: 126.18
Flash Point: 110°(230°F)
Odor: Unpleasant
Boiling Point: 256°
Beilstein Reference: 2039306
UN No.: 1759
MDL No.: MFCD00004850
Formula:
C₆H₆OS Boiling Pt: 256 °C Melting Pt: 33…37 °C Density: 1.255 Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004850 CAS Number: 637-89-8 EINECS: 211-307-6 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...