7-Hydroxycoumarin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone

L04082-14 L04082-22
AAAL04082-14EA 73.83 USD
AAAL04082-14 AAAL04082-22
7-Hydroxycoumarin 98%
7-Hydroxycoumarin

Purity: 98% CAS No.: 93-35-6 Molecular Formula: C9H6O3 Formula Weight: 162.14 Melting Point: 228-232° Beilstein Reference: 127683 Merck Reference: 14,9847 MDL No.: MFCD00006878

Formula: C₉H₆O₃
MW: 162.14 g/mol
Boiling Pt: 382
Melting Pt: 228…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00006878
CAS Number: 93-35-6
EINECS: 202-240-3
Merck Index: 14,9847

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR