o-Cresolphthalein complexone indicator for metal titration
Supplier: Thermo Scientific Chemicals
L03859-06
L03859-14
L03859-03
AAAL03859-06EA
63.57
USD
AAAL03859-06
AAAL03859-14
AAAL03859-03
o-Cresolphthalein complexone indicator for metal titration
o-Cresolphthalein complexone
CAS No.: 2411-89-4
Molecular Formula: C32H32N2O12
Formula Weight: 636.62
Melting Point: ca 198° dec.
Beilstein Reference: 381994
MDL No.: MFCD00005911
Formula:
C₃₂H₃₂N₂O₁₂ Melting Pt: ca 198 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005911 CAS Number: 2411-89-4 EINECS: 219-318-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...