Berberine chloride hydrate, (∼17% H₂O) 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Natural Yellow 18

L03807-06
AAAL03807-06EA 32.5 USD
AAAL03807-06
Berberine chloride hydrate, (∼17% H₂O) 96%
Berberine chloride hydrate

Antiprotozoal and antidiarrheal agent. Apoptosis inhibitor


Purity: 97% CAS No.: 141433-60-5 Molecular Formula: C20H18ClNO4.xH2O Formula Weight: 371.82(anhy) Melting Point: ca 200° dec. Beilstein Reference: 3836585 MDL No.: MFCD00149998

Formula: C₂₀H₁₈ClNO₄
MW: 371.82 g/mol
Melting Pt: ∼ 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011939
CAS Number: 141433-60-5
EINECS: 211-195-9
Merck Index: 14,1154

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR