Neopentyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane

L03579-14 L03579-06
AAAL03579-14EA 230 USD
AAAL03579-14 AAAL03579-06
Neopentyl bromide 98%
Neopentyl bromide

Purity: 98% CAS No.: 630-17-1 Molecular Formula: (CH3)3CCH2Br Formula Weight: 151.05 Flash Point: 6°(42°F) Refractive Index: 1.4360 Boiling Point: 104-106° Beilstein Reference: 1730989 UN No.: 1993 MDL No.: MFCD00000209

Formula: C₅H₁₁Br
Boiling Pt: 104…106 °C
Density: 1.195
Flash Pt: 6 °C (42 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000209
CAS Number: 630-17-1
EINECS: 211-132-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR