Neopentyl bromide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane
L03579-14
L03579-06
AAAL03579-14EA
230
USD
AAAL03579-14
AAAL03579-06
Neopentyl bromide 98%
Neopentyl bromide
Purity: 98%
CAS No.: 630-17-1
Molecular Formula: (CH3)3CCH2Br
Formula Weight: 151.05
Flash Point: 6°(42°F)
Refractive Index: 1.4360
Boiling Point: 104-106°
Beilstein Reference: 1730989
UN No.: 1993
MDL No.: MFCD00000209
Formula:
C₅H₁₁Br Boiling Pt: 104…106 °C Density: 1.195 Flash Pt: 6 °C (42 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000209 CAS Number: 630-17-1 EINECS: 211-132-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...