1-Naphthylmethylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Naphthalenemethylamine, Naphthalen-1-ylmethanamine, Naphthalene-1-methylamine, 1-(Aminomethyl)naphthalene

L03541-06 L03541-14 L03541-03
AAAL03541-06EA 84.8 USD
AAAL03541-06 AAAL03541-14 AAAL03541-03
1-Naphthylmethylamine 98+%
1-Naphthylmethylamine

Purity: 98+% CAS No.: 118-31-0 Molecular Formula: C11H11N Formula Weight: 157.22 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.6430 Boiling Point: 289-292° Beilstein Reference: 2206459 UN No.: 2735 MDL No.: MFCD00004048

Formula: C₁₁H₁₁N
MW: 157.22 g/mol
Boiling Pt: 92
Melting Pt: 267…269 °C
Density: 1.075
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004048
CAS Number: 118-31-0
EINECS: 204-244-0
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR